Product Details

SNP ID
rs115390619
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:47610888 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTTCCCATGCACTGGTAGGACTT[C/T]TGGGACATGTTGACCTCCCTTGCCT
Phenotype
MIM: 605690
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLTSCR1 PubMed Links

Gene Details

Gene
GLTSCR1
Gene Name
glioma tumor suppressor candidate region gene 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015711.3 Intron NP_056526.3
XM_005258833.4 Intron XP_005258890.1
XM_006723180.3 Intron XP_006723243.1
XM_011526882.2 Intron XP_011525184.1
XM_011526883.2 Intron XP_011525185.1

View Full Product Details