Product Details

SNP ID
rs116718228
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55477882 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCCAGACATTATTGTAGGCACT[A/G]GGATGGCATACAACAGTGAACCAGA
Phenotype
MIM: 610671
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NAT14 PubMed Links

Gene Details

Gene
NAT14
Gene Name
N-acetyltransferase 14 (putative)
There are no transcripts associated with this gene.

Gene
ZNF628
Gene Name
zinc finger protein 628
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033113.2 Intron NP_149104.3
XM_005259371.3 Intron XP_005259428.1

View Full Product Details