Product Details

SNP ID
rs116806392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:34434847 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATTTTTTTTTTCCCAAAAACTCA[C/T]ACTGTTTGTTTTACTTCCAGCTGCC
Phenotype
MIM: 615661 MIM: 613295
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDCD2L PubMed Links

Gene Details

Gene
PDCD2L
Gene Name
programmed cell death 2 like
There are no transcripts associated with this gene.

Gene
UBA2
Gene Name
ubiquitin like modifier activating enzyme 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005499.2 Intron NP_005490.1
XM_005258404.3 Intron XP_005258461.2
XM_006722962.2 Intron XP_006723025.1
XM_011526304.2 Intron XP_011524606.1
XM_017026134.1 Intron XP_016881623.1

View Full Product Details