Product Details

SNP ID
rs113534319
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:501843 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCCCAGCAGGGCTCCACACACA[A/C]CCCCAGGAGCCCAGGCTCCACCAGG
Phenotype
MIM: 102670
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MADCAM1 PubMed Links

Gene Details

Gene
MADCAM1
Gene Name
mucosal vascular addressin cell adhesion molecule 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_130760.2 852 Missense Mutation AAC,ACC N281T NP_570116.2
NM_130762.2 852 Intron NP_570118.1
Gene
TPGS1
Gene Name
tubulin polyglutamylase complex subunit 1
There are no transcripts associated with this gene.

View Full Product Details