Product Details

SNP ID
rs116790932
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:5545528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGTTTGGCAGAGAGAAAGTGACT[A/G]TGGGAGGAGAGGGCTCCCAGGCAGA
Phenotype
MIM: 614124
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPCPD1 PubMed Links

Gene Details

Gene
GPCPD1
Gene Name
glycerophosphocholine phosphodiesterase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019593.3 4516 UTR 3 NP_062539.1
XM_005260758.4 4516 UTR 3 XP_005260815.1
XM_005260761.1 4516 UTR 3 XP_005260818.1
XM_006723596.3 4516 UTR 3 XP_006723659.1

View Full Product Details