Product Details

SNP ID
rs113275808
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:55636454 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACATTGAGTGACATCTAAACTAA[C/T]AGACATTTAAATTTACATGGTCCAA
Phenotype
MIM: 610316
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PNPT1 PubMed Links

Gene Details

Gene
PNPT1
Gene Name
polyribonucleotide nucleotidyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033109.4 Intron NP_149100.2
XM_005264629.2 Intron XP_005264686.1
XM_017005172.1 Intron XP_016860661.1

View Full Product Details