Product Details

SNP ID
rs116736857
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:62200269 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTAAGAATGTTACACATTATTTT[C/T]TGTGCATCTATAACTTAAAAAAAAC
Phenotype
MIM: 605581
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
B3GNT2 PubMed Links

Gene Details

Gene
B3GNT2
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319075.1 Intron NP_001306004.1
NM_006577.5 Intron NP_006568.2
Gene
MIR5192
Gene Name
microRNA 5192
There are no transcripts associated with this gene.

View Full Product Details