Product Details

SNP ID
rs114167904
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:57627782 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGCATCAATTCCTTCTAACCATA[A/G]GACAGTTCAGTCTTTCTGTTCTTAA
Phenotype
MIM: 616519
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DENND6A PubMed Links

Gene Details

Gene
DENND6A
Gene Name
DENN domain containing 6A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152678.2 2330 UTR 3 NP_689891.1
XM_006713019.3 2330 UTR 3 XP_006713082.1
XM_006713020.2 2330 UTR 3 XP_006713083.1
XM_017005864.1 2330 UTR 3 XP_016861353.1
Gene
PDE12
Gene Name
phosphodiesterase 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322176.1 2330 Intron NP_001309105.1
NM_001322177.1 2330 Intron NP_001309106.1
NM_177966.6 2330 Intron NP_808881.3

View Full Product Details