Product Details

SNP ID
rs113807887
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:169702537 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTAGCCTCGCAGGTCCTGTCTCC[C/G]GGAAGCTGCGCGGACGAGGAGGAGG
Phenotype
MIM: 616987 MIM: 613069
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C6orf120 PubMed Links

Gene Details

Gene
C6orf120
Gene Name
chromosome 6 open reading frame 120
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029863.2 426 Silent Mutation CCC,CCG P26P NP_001025034.1
NM_001317342.1 426 Silent Mutation CCC,CCG P45P NP_001304271.1
Gene
PHF10
Gene Name
PHD finger protein 10
There are no transcripts associated with this gene.

Gene
WDR27
Gene Name
WD repeat domain 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202550.1 426 Intron NP_001189479.1
NM_182552.4 426 Intron NP_872358.4
XM_011535682.2 426 Intron XP_011533984.1
XM_011535684.2 426 Intron XP_011533986.1
XM_011535685.2 426 Intron XP_011533987.1
XM_011535687.2 426 Intron XP_011533989.1
XM_011535688.2 426 Intron XP_011533990.1
XM_011535689.2 426 Intron XP_011533991.1
XM_011535690.2 426 Intron XP_011533992.1
XM_011535691.2 426 Intron XP_011533993.1
XM_011535692.2 426 Intron XP_011533994.1
XM_011535693.2 426 Intron XP_011533995.1
XM_011535694.2 426 Intron XP_011533996.1
XM_011535696.2 426 Intron XP_011533998.1
XM_011535697.2 426 Intron XP_011533999.1
XM_017010658.1 426 Intron XP_016866147.1
XM_017010659.1 426 Intron XP_016866148.1
XM_017010660.1 426 Intron XP_016866149.1
XM_017010661.1 426 Intron XP_016866150.1
XM_017010662.1 426 Intron XP_016866151.1
XM_017010663.1 426 Intron XP_016866152.1
XM_017010664.1 426 Intron XP_016866153.1
XM_017010665.1 426 Intron XP_016866154.1
XM_017010666.1 426 Intron XP_016866155.1
XM_017010667.1 426 Intron XP_016866156.1
XM_017010668.1 426 Intron XP_016866157.1
XM_017010669.1 426 Intron XP_016866158.1
XM_017010670.1 426 Intron XP_016866159.1
XM_017010671.1 426 Intron XP_016866160.1
XM_017010672.1 426 Intron XP_016866161.1
XM_017010673.1 426 Intron XP_016866162.1
XM_017010674.1 426 Intron XP_016866163.1
XM_017010675.1 426 Intron XP_016866164.1

View Full Product Details