Product Details

SNP ID
rs116084833
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:169702877 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGTGTACTACGACGGCACGGTC[A/G]AGCAGCACCCGTTCGGCGAGGCCGC
Phenotype
MIM: 616987 MIM: 613069
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C6orf120 PubMed Links

Gene Details

Gene
C6orf120
Gene Name
chromosome 6 open reading frame 120
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029863.2 766 Missense Mutation AAG,GAG K140E NP_001025034.1
NM_001317342.1 766 Missense Mutation AAG,GAG K159E NP_001304271.1
Gene
PHF10
Gene Name
PHD finger protein 10
There are no transcripts associated with this gene.

Gene
WDR27
Gene Name
WD repeat domain 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202550.1 766 Intron NP_001189479.1
NM_182552.4 766 Intron NP_872358.4
XM_011535682.2 766 Intron XP_011533984.1
XM_011535684.2 766 Intron XP_011533986.1
XM_011535685.2 766 Intron XP_011533987.1
XM_011535687.2 766 Intron XP_011533989.1
XM_011535688.2 766 Intron XP_011533990.1
XM_011535689.2 766 Intron XP_011533991.1
XM_011535690.2 766 Intron XP_011533992.1
XM_011535691.2 766 Intron XP_011533993.1
XM_011535692.2 766 Intron XP_011533994.1
XM_011535693.2 766 Intron XP_011533995.1
XM_011535694.2 766 Intron XP_011533996.1
XM_011535696.2 766 Intron XP_011533998.1
XM_011535697.2 766 Intron XP_011533999.1
XM_017010658.1 766 Intron XP_016866147.1
XM_017010659.1 766 Intron XP_016866148.1
XM_017010660.1 766 Intron XP_016866149.1
XM_017010661.1 766 Intron XP_016866150.1
XM_017010662.1 766 Intron XP_016866151.1
XM_017010663.1 766 Intron XP_016866152.1
XM_017010664.1 766 Intron XP_016866153.1
XM_017010665.1 766 Intron XP_016866154.1
XM_017010666.1 766 Intron XP_016866155.1
XM_017010667.1 766 Intron XP_016866156.1
XM_017010668.1 766 Intron XP_016866157.1
XM_017010669.1 766 Intron XP_016866158.1
XM_017010670.1 766 Intron XP_016866159.1
XM_017010671.1 766 Intron XP_016866160.1
XM_017010672.1 766 Intron XP_016866161.1
XM_017010673.1 766 Intron XP_016866162.1
XM_017010674.1 766 Intron XP_016866163.1
XM_017010675.1 766 Intron XP_016866164.1

View Full Product Details