Product Details

SNP ID
rs115254671
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:44903212 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATTTAATAATTATTATAGTCCTC[C/T]CCAAGAAATTCCACATGGAAACTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C14orf28 PubMed Links

Gene Details

Gene
C14orf28
Gene Name
chromosome 14 open reading frame 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017923.1 796 Missense Mutation CCC,CTC P177L NP_001017923.1
XM_011536408.1 796 UTR 5 XP_011534710.1
Gene
LOC101927418
Gene Name
uncharacterized LOC101927418
There are no transcripts associated with this gene.

View Full Product Details