Product Details

SNP ID
rs117363681
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:49986908 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATTTCAGATGAGAATGGACGTA[C/T]CCTCTCCAGAGGCAAAGAAGTTTGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC01588 PubMed Links

Gene Details

Gene
LINC01588
Gene Name
long intergenic non-protein coding RNA 1588
There are no transcripts associated with this gene.

View Full Product Details