Product Details

SNP ID
rs116218309
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:114471691 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACAGCTGCTGCCAATGGTCATTT[A/C]CTATTGGTTGGTATTGAGCCTTGGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NXPE2 PubMed Links

Gene Details

Gene
NXPE2
Gene Name
neurexophilin and PC-esterase domain family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182495.5 Intron NP_872301.2
XM_011542604.2 Intron XP_011540906.1
XM_017017206.1 Intron XP_016872695.1
XM_017017207.1 Intron XP_016872696.1
XM_017017208.1 Intron XP_016872697.1
XM_017017209.1 Intron XP_016872698.1
XM_017017210.1 Intron XP_016872699.1
XM_017017211.1 Intron XP_016872700.1
XM_017017212.1 Intron XP_016872701.1

View Full Product Details