Product Details

SNP ID
rs117780711
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:132950380 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAAATGCATACCTCCATTCCTTC[C/T]ACCACACCGCCTGCTTACTCAGGTT
Phenotype
MIM: 604383 MIM: 605284
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GFI1B PubMed Links

Gene Details

Gene
GFI1B
Gene Name
growth factor independent 1B transcriptional repressor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135031.1 Intron NP_001128503.1
NM_004188.6 Intron NP_004179.3
XM_006717297.3 Intron XP_006717360.1
XM_011519068.2 Intron XP_011517370.1
XM_011519069.2 Intron XP_011517371.1
XM_011519070.2 Intron XP_011517372.1
XM_017015175.1 Intron XP_016870664.1
XM_017015176.1 Intron XP_016870665.1
Gene
MIR548AW
Gene Name
microRNA 548aw
There are no transcripts associated with this gene.

Gene
TSC1
Gene Name
tuberous sclerosis 1
There are no transcripts associated with this gene.

View Full Product Details