Product Details

SNP ID
rs114581152
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:1122689 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGGCTGCTGCATTATTAGAGAA[C/T]GCCTATGCCAAAATGGGAGGAGGGA
Phenotype
MIM: 612041
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105374344 PubMed Links

Gene Details

Gene
LOC105374344
Gene Name
uncharacterized LOC105374344
There are no transcripts associated with this gene.

Gene
RNF212
Gene Name
ring finger protein 212
There are no transcripts associated with this gene.

Gene
TMED11P
Gene Name
transmembrane p24 trafficking protein 11, pseudogene
There are no transcripts associated with this gene.

View Full Product Details