Product Details

SNP ID
rs142238416
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:109869964 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATAAATCCTCACCTTGGTCTTCA[A/C]AGGAACCACAAGGACAACATTCTCA
Phenotype
MIM: 602443
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
XPNPEP1 PubMed Links

Gene Details

Gene
XPNPEP1
Gene Name
X-prolyl aminopeptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167604.1 1729 Missense Mutation GTG,TTG V564L NP_001161076.1
NM_001324128.1 1729 Missense Mutation GTG,TTG V474L NP_001311057.1
NM_001324131.1 1729 Missense Mutation GTG,TTG V457L NP_001311060.1
NM_001324132.1 1729 Missense Mutation GTG,TTG V545L NP_001311061.1
NM_001324133.1 1729 Missense Mutation GTG,TTG V588L NP_001311062.1
NM_001324134.1 1729 Missense Mutation GTG,TTG V414L NP_001311063.1
NM_001324135.1 1729 Missense Mutation GTG,TTG V571L NP_001311064.1
NM_001324136.1 1729 Missense Mutation GTG,TTG V583L NP_001311065.1
NM_020383.3 1729 Missense Mutation GTG,TTG V588L NP_065116.3
XM_011540134.1 1729 Missense Mutation GTG,TTG V530L XP_011538436.1
XM_017016613.1 1729 Missense Mutation GTG,TTG V540L XP_016872102.1
XM_017016614.1 1729 Missense Mutation GTG,TTG V474L XP_016872103.1
XM_017016615.1 1729 Missense Mutation GTG,TTG V474L XP_016872104.1

View Full Product Details