Product Details

SNP ID
hCV160658817
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:46036858 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGAGCCACGCTCCTATGTGAGGG[C/A]GGGACCAGTCACCAGTCCTTCACCT
Phenotype
MIM: 157145 MIM: 601984
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
MSMB PubMed Links

Gene Details

Gene
MSMB
Gene Name
microseminoprotein beta
There are no transcripts associated with this gene.

Gene
NCOA4
Gene Name
nuclear receptor coactivator 4
There are no transcripts associated with this gene.

View Full Product Details