Product Details

SNP ID
rs147836406
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:22536947 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGACTTTTAAAATGCACACGCGC[A/G]CACACACTCACCCCCCCCCAACACA
Phenotype
MIM: 603140
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PIP4K2A PubMed Links

Gene Details

Gene
PIP4K2A
Gene Name
phosphatidylinositol-5-phosphate 4-kinase type 2 alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005028.4 1365 UTR 3 NP_005019.2
XM_006717450.2 1365 UTR 3 XP_006717513.1
XM_011519496.1 1365 UTR 3 XP_011517798.1
XM_017016330.1 1365 UTR 3 XP_016871819.1
XM_017016331.1 1365 UTR 3 XP_016871820.1
XM_017016332.1 1365 UTR 3 XP_016871821.1

View Full Product Details