Product Details

SNP ID
rs141060948
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:69006018 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCCGTGTTCAGGATGGAGAACA[A/C]CGCCTTGCTGAAGAGGTAGCCCACA
Phenotype
MIM: 607233
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MRGPRF PubMed Links

Gene Details

Gene
MRGPRF
Gene Name
MAS related GPR family member F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098515.1 1157 Missense Mutation GTG,TTG V98L NP_001091985.1
NM_145015.4 1157 Missense Mutation GTG,TTG V98L NP_659452.3
XM_017017170.1 1157 Missense Mutation GTG,TTG V98L XP_016872659.1
Gene
MRGPRF-AS1
Gene Name
MRGPRF antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details