Product Details
- SNP ID
-
rs141683852
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:5856835 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCATAGATTACAGGATTGAGGGCTG[A/G]TGGGACAACCACATACAGGTTGGCT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR52E8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs61059077] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR52E8
- Gene Name
- olfactory receptor family 52 subfamily E member 8
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001005168.1 |
868 |
Missense Mutation |
CCA,TCA |
P290S |
NP_001005168.1 |
View Full Product Details