Product Details

SNP ID
rs141816603
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4914965 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGAATCTCTCAGCCCTGGAGGC[A/G]ATGGACAGCACGGTGCGCAGGATCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR51A7 PubMed Links

Gene Details

Gene
OR51A7
Gene Name
olfactory receptor family 51 subfamily A member 7
There are no transcripts associated with this gene.

Gene
OR51G1
Gene Name
olfactory receptor family 51 subfamily G member 1 (gene/pseudogene)
There are no transcripts associated with this gene.

Gene
OR51G2
Gene Name
olfactory receptor family 51 subfamily G member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005238.1 699 Silent Mutation ATC,ATT I233I NP_001005238.1

View Full Product Details