Product Details

SNP ID
rs142707103
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63912698 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCTTGAACAGCCAAAAGCTGCTC[A/G]TCTGTGGTCCAGCGGGAGTTGAACT
Phenotype
MIM: 600526 MIM: 616019
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MARK2 PubMed Links

Gene Details

Gene
MARK2
Gene Name
microtubule affinity regulating kinase 2
There are no transcripts associated with this gene.

Gene
RCOR2
Gene Name
REST corepressor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173587.3 1393 Silent Mutation GAC,GAT D335D NP_775858.2
XM_005273932.3 1393 Intron XP_005273989.1

View Full Product Details