Product Details
- SNP ID
-
rs145474169
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:47472344 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGCTTGGGCCGAAACAGGATTGTC[A/G]TAACTTACAAAACCTGTGTGTCCAA
- Phenotype
-
MIM: 601074
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CELF1
PubMed Links
Gene Details
- Gene
- CELF1
- Gene Name
- CUGBP, Elav-like family member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001025596.2 |
1609 |
Silent Mutation |
TAC,TAT |
Y449Y |
NP_001020767.1 |
NM_001172639.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y475Y |
NP_001166110.1 |
NM_001172640.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y448Y |
NP_001166111.1 |
NM_006560.3 |
1609 |
Silent Mutation |
TAC,TAT |
Y445Y |
NP_006551.1 |
NM_198700.2 |
1609 |
Silent Mutation |
TAC,TAT |
Y446Y |
NP_941989.1 |
XM_011519848.2 |
1609 |
Silent Mutation |
TAC,TAT |
Y450Y |
XP_011518150.1 |
XM_011519850.2 |
1609 |
Silent Mutation |
TAC,TAT |
Y476Y |
XP_011518152.2 |
XM_011519852.2 |
1609 |
Silent Mutation |
TAC,TAT |
Y477Y |
XP_011518154.1 |
XM_011519854.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y450Y |
XP_011518156.1 |
XM_011519855.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y450Y |
XP_011518157.1 |
XM_011519856.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y450Y |
XP_011518158.1 |
XM_011519857.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y450Y |
XP_011518159.1 |
XM_011519858.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y450Y |
XP_011518160.1 |
XM_011519859.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y446Y |
XP_011518161.1 |
XM_017017101.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y477Y |
XP_016872590.1 |
XM_017017102.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y477Y |
XP_016872591.1 |
XM_017017103.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y477Y |
XP_016872592.1 |
XM_017017104.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y477Y |
XP_016872593.1 |
XM_017017105.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y476Y |
XP_016872594.1 |
XM_017017106.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y476Y |
XP_016872595.1 |
XM_017017107.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y476Y |
XP_016872596.1 |
XM_017017108.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y473Y |
XP_016872597.1 |
XM_017017109.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y473Y |
XP_016872598.1 |
XM_017017110.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y473Y |
XP_016872599.1 |
XM_017017111.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y472Y |
XP_016872600.1 |
XM_017017112.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y472Y |
XP_016872601.1 |
XM_017017113.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y472Y |
XP_016872602.1 |
XM_017017114.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y472Y |
XP_016872603.1 |
XM_017017115.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y450Y |
XP_016872604.1 |
XM_017017116.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y450Y |
XP_016872605.1 |
XM_017017117.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y450Y |
XP_016872606.1 |
XM_017017118.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y450Y |
XP_016872607.1 |
XM_017017119.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y450Y |
XP_016872608.1 |
XM_017017120.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y450Y |
XP_016872609.1 |
XM_017017121.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y450Y |
XP_016872610.1 |
XM_017017122.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y450Y |
XP_016872611.1 |
XM_017017123.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y450Y |
XP_016872612.1 |
XM_017017124.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y449Y |
XP_016872613.1 |
XM_017017125.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y449Y |
XP_016872614.1 |
XM_017017126.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y449Y |
XP_016872615.1 |
XM_017017127.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y449Y |
XP_016872616.1 |
XM_017017128.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y449Y |
XP_016872617.1 |
XM_017017129.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y449Y |
XP_016872618.1 |
XM_017017130.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y449Y |
XP_016872619.1 |
XM_017017131.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y449Y |
XP_016872620.1 |
XM_017017132.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y449Y |
XP_016872621.1 |
XM_017017133.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y446Y |
XP_016872622.1 |
XM_017017134.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y446Y |
XP_016872623.1 |
XM_017017135.1 |
1609 |
Silent Mutation |
TAC,TAT |
Y445Y |
XP_016872624.1 |
View Full Product Details