Product Details

SNP ID
rs145616541
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:1276214 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCACAGTGCTGCCTCTATCACCTC[C/T]CACCCCAGCCTCGCAAGGCCTGTTG
Phenotype
MIM: 606277
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TOLLIP PubMed Links

Gene Details

Gene
TOLLIP
Gene Name
toll interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318512.1 1814 UTR 3 NP_001305441.1
NM_001318514.1 1814 UTR 3 NP_001305443.1
NM_001318515.1 1814 UTR 3 NP_001305444.1
NM_001318516.1 1814 UTR 3 NP_001305445.1
NM_019009.3 1814 UTR 3 NP_061882.2
XM_017017931.1 1814 UTR 3 XP_016873420.1

View Full Product Details