Product Details

SNP ID
rs146937207
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66278477 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCATGGCGTTCACCTTCGCCGC[A/G]TTCTGCTACATGCTCACCCTGGTGC
Phenotype
MIM: 611288 MIM: 612565 MIM: 611484
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNIH2 PubMed Links

Gene Details

Gene
CNIH2
Gene Name
cornichon family AMPA receptor auxiliary protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182553.2 277 Silent Mutation GCA,GCG A7A NP_872359.1
Gene
RAB1B
Gene Name
RAB1B, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030981.2 277 Intron NP_112243.1
XM_017018378.1 277 Intron XP_016873867.1
Gene
YIF1A
Gene Name
Yip1 interacting factor homolog A, membrane trafficking protein
There are no transcripts associated with this gene.

View Full Product Details