Product Details

SNP ID
rs147186459
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59443344 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGATCAGAGGTGACACCCATCTGC[A/G]CACACCCATGTACTTCTTCCTAAGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR5A1 PubMed Links

Gene Details

Gene
OR5A1
Gene Name
olfactory receptor family 5 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004728.1 176 Missense Mutation CAC,CGC H59R NP_001004728.1

View Full Product Details