Product Details

SNP ID
rs147748696
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:36592778 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGCTGACAGATGGATGAGTGTG[C/T]GTTCTGCCAGATCCATGCACTGAGC
Phenotype
MIM: 179616
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C11orf74 PubMed Links

Gene Details

Gene
C11orf74
Gene Name
chromosome 11 open reading frame 74
There are no transcripts associated with this gene.

Gene
RAG2
Gene Name
recombination activating gene 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000536.3 1596 Missense Mutation CCC,CTC P464L NP_000527.2
NM_001243785.1 1596 Missense Mutation CCC,CTC P464L NP_001230714.1
NM_001243786.1 1596 Missense Mutation CCC,CTC P464L NP_001230715.1

View Full Product Details