Product Details

SNP ID
rs149135276
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4640227 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCATTTGCCACCCATTGCGCCATG[C/T]TTCTGTGCTGACAGGGTGTACTGTG
Phenotype
MIM: 611267
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR51D1 PubMed Links

Gene Details

Gene
OR51D1
Gene Name
olfactory receptor family 51 subfamily D member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004751.2 513 Missense Mutation GCT,GTT A146V NP_001004751.1
Gene
OR51E1
Gene Name
olfactory receptor family 51 subfamily E member 1
There are no transcripts associated with this gene.

View Full Product Details