Product Details

SNP ID
rs149330601
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:95790577 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCAAGCCCAGCACCAGCACCCTT[A/G]CCCTTTTCCATCAGGGGTTCAGCCT
Phenotype
MIM: 607951
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEP57 PubMed Links

Gene Details

Gene
CEP57
Gene Name
centrosomal protein 57
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243776.1 117 UTR 5 NP_001230705.1
NM_001243777.1 117 UTR 5 NP_001230706.1
NM_014679.4 117 UTR 5 NP_055494.2
XM_006718945.2 117 UTR 5 XP_006719008.1
XM_006718946.2 117 UTR 5 XP_006719009.1
XM_017018592.1 117 Intron XP_016874081.1
XM_017018593.1 117 UTR 5 XP_016874082.1
XM_017018594.1 117 UTR 5 XP_016874083.1
Gene
FAM76B
Gene Name
family with sequence similarity 76 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144664.4 117 Intron NP_653265.3
XM_005273775.4 117 Intron XP_005273832.1
XM_005273776.4 117 Intron XP_005273833.1
XM_011542610.2 117 Intron XP_011540912.1
XM_011542611.2 117 Intron XP_011540913.1
XM_011542612.1 117 Intron XP_011540914.1
XM_011542613.1 117 Intron XP_011540915.1

View Full Product Details