Product Details

SNP ID
rs150525831
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4821890 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCATGTTGATAAGAAATGTTGCC[A/G]TCATGTTGCCAGTCATGCTCTTTGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR51F2 PubMed Links

Gene Details

Gene
OR51F2
Gene Name
olfactory receptor family 51 subfamily F member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004753.1 505 Missense Mutation ATC,GTC I169V NP_001004753.1

View Full Product Details