Product Details

SNP ID
rs150605099
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124570223 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTTTCTCCTCTTCCTCGGGATCT[A/G]CTTGGTCACCATCGTGGGGAACCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR8A1 PubMed Links

Gene Details

Gene
OR8A1
Gene Name
olfactory receptor family 8 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005194.1 155 Missense Mutation TAC,TGC Y52C NP_001005194.1

View Full Product Details