Product Details

SNP ID
rs139486404
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:51913124 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGACCCTGTGAAGCCGTCTCGGGG[C/T]CCGCTGGTGACCTGCACGTGTGAGA
Phenotype
MIM: 601284
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACVRL1 PubMed Links

Gene Details

Gene
ACVRL1
Gene Name
activin A receptor like type 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000020.2 233 Silent Mutation GGC,GGT G29G NP_000011.2
NM_001077401.1 233 Silent Mutation GGC,GGT G29G NP_001070869.1
XM_005269235.2 233 Silent Mutation GGC,GGT G29G XP_005269292.1
XM_011539008.1 233 Silent Mutation GGC,GGT G43G XP_011537310.1

View Full Product Details