Product Details

SNP ID
rs139991199
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:68689593 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACCAAGAAACCAGGTTATCCCTC[A/G]AACTCCTAGCTCATTTCGACAGCCT
Phenotype
MIM: 607617
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100507250 PubMed Links

Gene Details

Gene
LOC100507250
Gene Name
uncharacterized LOC100507250
There are no transcripts associated with this gene.

Gene
NUP107
Gene Name
nucleoporin 107
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005269037.3 267 Missense Mutation CAA,CGA Q54R XP_005269094.1
XM_011538576.1 267 Missense Mutation AAA,GAA K16E XP_011536878.1

View Full Product Details