Product Details

SNP ID
rs143099272
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:14882240 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGTCTGTAGTCATCACAGGCTT[C/G]CCTATTGAGCTCGTGGACAGGCTTA
Phenotype
MIM: 154870
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MGP PubMed Links

Gene Details

Gene
MGP
Gene Name
matrix Gla protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000900.3 414 Missense Mutation CAA,GAA Q71E NP_000891.2
NM_001190839.1 414 Missense Mutation CAA,GAA Q96E NP_001177768.1

View Full Product Details