Product Details

SNP ID
rs143775343
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:123415906 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCTTGGATCTAGGTCTGTTTCCC[C/T]GATCGAAAAAAGAACCTGGTGTGGA
Phenotype
MIM: 607240 MIM: 614093
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KMT5A PubMed Links

Gene Details

Gene
KMT5A
Gene Name
lysine methyltransferase 5A
There are no transcripts associated with this gene.

Gene
RILPL2
Gene Name
Rab interacting lysosomal protein like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145058.2 934 Silent Mutation NP_659495.1
XM_011538012.2 934 Intron XP_011536314.1
XM_017018959.1 934 Intron XP_016874448.1

View Full Product Details