Product Details

SNP ID
rs146057190
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:122172651 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTTGAGATATGCCCGGATGGCTG[G/T]GCTGTGGATGTTGTTTGGCGGCTGG
Phenotype
MIM: 609509
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IL31 PubMed Links

Gene Details

Gene
IL31
Gene Name
interleukin 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014336.1 283 Missense Mutation ACA,CCA T86P NP_001014358.1
Gene
LRRC43
Gene Name
leucine rich repeat containing 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098519.1 283 Intron NP_001091989.1
NM_152759.4 283 Intron NP_689972.3
XM_017019124.1 283 Intron XP_016874613.1
XM_017019125.1 283 Intron XP_016874614.1

View Full Product Details