Product Details

SNP ID
rs146125088
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:123423052 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAAGGTGACACTTACAGCTTTTT[A/T]ATGATTGTCTTCTCCTCCTTGTTCC
Phenotype
MIM: 614093
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RILPL2 PubMed Links

Gene Details

Gene
RILPL2
Gene Name
Rab interacting lysosomal protein like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145058.2 854 Silent Mutation ATA,ATT I199I NP_659495.1
XM_011538012.2 854 Silent Mutation ATA,ATT I199I XP_011536314.1
XM_017018959.1 854 Silent Mutation ATA,ATT I199I XP_016874448.1

View Full Product Details