Product Details

SNP ID
rs146368046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:110486558 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGTCCTCACTCATCACAGAAGCC[A/G]CCTTAGCTCCCGTTACTCACAGAAA
Phenotype
MIM: 606932
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM216A PubMed Links

Gene Details

Gene
FAM216A
Gene Name
family with sequence similarity 216 member A
There are no transcripts associated with this gene.

Gene
VPS29
Gene Name
VPS29, retromer complex component
There are no transcripts associated with this gene.

View Full Product Details