Product Details

SNP ID
rs150132451
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48149685 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCGGGAGATGGGTGTCTGGCCT[A/G]TTAGGTTGATGACTCTGACCTCTGC
Phenotype
MIM: 615053 MIM: 610681
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASB8 PubMed Links

Gene Details

Gene
ASB8
Gene Name
ankyrin repeat and SOCS box containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319296.1 772 Missense Mutation ACA,ATA T183I NP_001306225.1
NM_001319297.1 772 Missense Mutation ACA,ATA T183I NP_001306226.1
NM_001319298.1 772 Missense Mutation ACA,ATA T122I NP_001306227.1
NM_001319299.1 772 Missense Mutation ACA,ATA T122I NP_001306228.1
NM_001319300.1 772 UTR 3 NP_001306229.1
NM_001319301.1 772 UTR 3 NP_001306230.1
NM_024095.4 772 Missense Mutation ACA,ATA T183I NP_077000.1
Gene
PFKM
Gene Name
phosphofructokinase, muscle
There are no transcripts associated with this gene.

View Full Product Details