Product Details

SNP ID
rs150694299
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:22055199 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATTGAGAATGTGGCCAAACAAT[A/T]TGGTGCACAAGTTCATCGAAGAAGT
Phenotype
MIM: 603316
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CMAS PubMed Links

Gene Details

Gene
CMAS
Gene Name
cytidine monophosphate N-acetylneuraminic acid synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018686.5 441 Missense Mutation TAT,TTT Y104F NP_061156.1

View Full Product Details