Product Details

SNP ID
rs146304026
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:51865467 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCTTGCCTGCTTCCGGTCCCTGG[C/T]GGCATCCTCTCCCAGTATTCGCCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC70 PubMed Links

Gene Details

Gene
CCDC70
Gene Name
coiled-coil domain containing 70
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031290.2 385 Missense Mutation GCG,GTG A30V NP_112580.2

View Full Product Details