Product Details

SNP ID
rs147499321
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:27254239 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTAGGAGTTGTTCCTTTGGCCAC[A/G]TATATGCGAATCTATAAGAAAGGTG
Phenotype
MIM: 603636
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPL21 PubMed Links

Gene Details

Gene
RPL21
Gene Name
ribosomal protein L21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000982.3 Intron NP_000973.2
Gene
SNORA27
Gene Name
small nucleolar RNA, H/ACA box 27
There are no transcripts associated with this gene.

Gene
SNORD102
Gene Name
small nucleolar RNA, C/D box 102
There are no transcripts associated with this gene.

View Full Product Details