Product Details

SNP ID
rs140518747
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:69570620 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCAGGAACAGCCTGGGCAGGCAC[A/G]TAAGGTAGCTCTGCTTCTTAAGGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC177 PubMed Links

Gene Details

Gene
CCDC177
Gene Name
coiled-coil domain containing 177
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271507.1 3373 UTR 3 NP_001258436.1

View Full Product Details