Product Details

SNP ID
rs141336782
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:69572758 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAACCAGGGTCAGGGCAGACGGGC[G/T]GCCCGGAGCGTTGGTGGTGGAGGAC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC177 PubMed Links

Gene Details

Gene
CCDC177
Gene Name
coiled-coil domain containing 177
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271507.1 1235 Missense Mutation AGC,CGC S289R NP_001258436.1

View Full Product Details