Product Details

SNP ID
rs142310777
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:105413567 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGGTGGTCCAGGATGCTGGTGAT[G/T]CTGGGATGGTTGCCCTCTCTGCTCC
Phenotype
MIM: 603526
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC100507437 PubMed Links

Gene Details

Gene
LOC100507437
Gene Name
uncharacterized LOC100507437
There are no transcripts associated with this gene.

Gene
MTA1
Gene Name
metastasis associated 1
There are no transcripts associated with this gene.

Gene
TEX22
Gene Name
testis expressed 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195082.1 2553 UTR 3 NP_001182011.1
XM_006720234.3 2553 UTR 3 XP_006720297.1

View Full Product Details