Product Details

SNP ID
rs143731263
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:58304979 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAGGATAATACCACACAATTGGT[A/G]CAAGATGACCAAGTAAAGGGTCCTT
Phenotype
MIM: 180201
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARID4A PubMed Links

Gene Details

Gene
ARID4A
Gene Name
AT-rich interaction domain 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002892.3 526 Silent Mutation GTA,GTG V51V NP_002883.3
NM_023000.2 526 Silent Mutation GTA,GTG V51V NP_075376.2
NM_023001.2 526 Silent Mutation GTA,GTG V51V NP_075377.2
XM_005267964.2 526 Silent Mutation GTA,GTG V51V XP_005268021.1
XM_005267965.2 526 Silent Mutation GTA,GTG V51V XP_005268022.1
XM_017021560.1 526 Silent Mutation GTA,GTG V51V XP_016877049.1
XM_017021561.1 526 Silent Mutation GTA,GTG V51V XP_016877050.1
XM_017021562.1 526 Silent Mutation GTA,GTG V51V XP_016877051.1
XM_017021563.1 526 Silent Mutation GTA,GTG V51V XP_016877052.1
XM_017021564.1 526 UTR 5 XP_016877053.1
XM_017021565.1 526 Intron XP_016877054.1
Gene
PSMA3-AS1
Gene Name
PSMA3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details