Product Details

SNP ID
rs147041367
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:72971017 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCGCAGGCCCGGCAGTGGTGCTT[C/G]GAGAGCTTGATGCTGAACTCCTTCC
Phenotype
MIM: 616372 MIM: 605471
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DCAF4 PubMed Links

Gene Details

Gene
DCAF4
Gene Name
DDB1 and CUL4 associated factor 4
There are no transcripts associated with this gene.

Gene
ZFYVE1
Gene Name
zinc finger FYVE-type containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281734.1 1056 Silent Mutation TCC,TCG S719S NP_001268663.1
NM_001281735.1 1056 Silent Mutation TCC,TCG S318S NP_001268664.1
NM_021260.3 1056 Silent Mutation TCC,TCG S733S NP_067083.1
NM_178441.2 1056 Silent Mutation TCC,TCG S318S NP_848535.1
XM_017021373.1 1056 Silent Mutation TCC,TCG S318S XP_016876862.1

View Full Product Details