Product Details

SNP ID
rs114352644
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:74923649 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTTCAGTGGTTTGTCGCTTACC[C/T]ATGCGGTTTACACTTTGTCAAGGCC
Phenotype
MIM: 603773
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COX5A PubMed Links

Gene Details

Gene
COX5A
Gene Name
cytochrome c oxidase subunit 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004255.3 Intron NP_004246.2

View Full Product Details