Product Details

SNP ID
rs115894895
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78961654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGGAGAAGCTGCTTTGGAAATT[A/G]ACATCACCATCAGCGGGAAGATTTG
Phenotype
MIM: 606600
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RASGRF1 PubMed Links

Gene Details

Gene
RASGRF1
Gene Name
Ras protein specific guanine nucleotide releasing factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145648.1 4469 UTR 3 NP_001139120.1
NM_002891.4 4469 UTR 3 NP_002882.3
NM_153815.2 4469 UTR 3 NP_722522.1
XM_011521866.2 4469 Intron XP_011520168.1
XM_011521867.2 4469 UTR 3 XP_011520169.1
XM_017022455.1 4469 UTR 3 XP_016877944.1
XM_017022456.1 4469 UTR 3 XP_016877945.1
XM_017022457.1 4469 Intron XP_016877946.1
XM_017022458.1 4469 Intron XP_016877947.1
XM_017022459.1 4469 UTR 3 XP_016877948.1

View Full Product Details