Product Details

SNP ID
rs117769984
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:49859724 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGTCTGAAAGCAACACAGTACA[C/T]GCTTTAAATTTTAAAAGTATACCTT
Phenotype
MIM: 609123
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP8B4 PubMed Links

Gene Details

Gene
ATP8B4
Gene Name
ATPase phospholipid transporting 8B4 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024837.3 4402 UTR 3 NP_079113.2
XM_011522046.2 4402 UTR 3 XP_011520348.1
XM_011522047.2 4402 UTR 3 XP_011520349.1
XM_011522048.1 4402 UTR 3 XP_011520350.1
XM_011522049.2 4402 UTR 3 XP_011520351.1
XM_011522051.2 4402 UTR 3 XP_011520353.1
XM_011522052.2 4402 UTR 3 XP_011520354.1
XM_011522053.1 4402 UTR 3 XP_011520355.1
XM_011522056.2 4402 UTR 3 XP_011520358.2
XM_011522058.2 4402 UTR 3 XP_011520360.1
XM_011522059.1 4402 UTR 3 XP_011520361.1
XM_011522060.1 4402 UTR 3 XP_011520362.1
XM_011522061.1 4402 UTR 3 XP_011520363.1
XM_011522062.1 4402 UTR 3 XP_011520364.1
XM_011522063.1 4402 UTR 3 XP_011520365.1
XM_011522064.1 4402 UTR 3 XP_011520366.1
XM_011522069.2 4402 UTR 3 XP_011520371.1
XM_011522070.1 4402 UTR 3 XP_011520372.1
XM_017022587.1 4402 UTR 3 XP_016878076.1
XM_017022588.1 4402 UTR 3 XP_016878077.1
XM_017022589.1 4402 UTR 3 XP_016878078.1
XM_017022590.1 4402 UTR 3 XP_016878079.1
XM_017022591.1 4402 UTR 3 XP_016878080.1
XM_017022592.1 4402 UTR 3 XP_016878081.1
XM_017022593.1 4402 Intron XP_016878082.1
XM_017022594.1 4402 UTR 3 XP_016878083.1
XM_017022595.1 4402 UTR 3 XP_016878084.1
XM_017022596.1 4402 UTR 3 XP_016878085.1
XM_017022597.1 4402 Intron XP_016878086.1
XM_017022598.1 4402 Intron XP_016878087.1
XM_017022599.1 4402 Intron XP_016878088.1

View Full Product Details